Assay Details
Gene Symbol: Traf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290413.1 | NP_001277342.1 | 8 - 9 | 1094 | 63 | |
NM_009422.3 | NP_033448.2 | 8 - 9 | 1073 | 63 | ||
GenBank mRNA | AF027570.1 | - | 7 - 8 | 985 | 63 | |
AK038136.1 | - | 8 - 9 | 1022 | 63 | ||
AK052934.1 | - | 8 - 9 | 1022 | 63 | ||
AK151653.1 | - | 8 - 9 | 1011 | 63 | ||
AK152174.1 | - | 8 - 9 | 1041 | 63 | ||
AK159581.1 | - | 8 - 9 | 1043 | 63 | ||
AK159585.1 | - | 1020 | 63 | |||
BC003801.1 | - | 7 - 8 | 999 | 3492839 | 63 | |
BC060625.1 | - | 6 - 7 | 3030 | 63 | ||
L35303.1 | - | 8 - 9 | 1010 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
