Assay Details
Gene Symbol: Gabra4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010251.2 | NP_034381.1 | 1 - 2 | 580 | 78 | |
GenBank mRNA | AK013727.1 | - | 1 - 2 | 581 | 78 | |
AK043806.1 | - | 1 - 2 | 198 | 78 | ||
AK137354.1 | - | 1 - 2 | 196 | 78 | ||
AK139970.1 | - | 1 - 2 | 254 | 78 | ||
AK141571.1 | - | 1 - 2 | 514 | 78 | ||
AK159299.1 | - | 1 - 2 | 540 | 78 | ||
AK162893.1 | - | 1 - 2 | 195 | 78 | ||
BC094603.1 | - | 1 - 2 | 208 | 6828002 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
