Assay Details
Gene Symbol: Gclc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010295.2 | NP_034425.1 | 3 - 4 | 723 | 66 | |
GenBank mRNA | AK144339.1 | - | 3 - 4 | 724 | 66 | |
AK160876.1 | - | 3 - 4 | 483 | 66 | ||
AK169676.1 | - | 3 - 4 | 696 | 66 | ||
BC019374.1 | - | 3 - 4 | 480 | 4195425 | 66 | |
U85414.1 | - | 3 - 4 | 448 | 66 | ||
U85498.1 | - | 3 - 4 | 448 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
