Assay Details
Gene Symbol: Igf1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010513.2 | NP_034643.2 | 10 - 11 | 2803 | 79 | |
GenBank mRNA | AF056187.1 | - | 10 - 11 | 2201 | 79 | |
AK137220.1 | - | 5 - 6 | 932 | 79 | ||
AK155884.1 | - | 9 - 10 | 1699 | 79 | ||
BC138868.1 | - | 10 - 11 | 2233 | 8861890 | 79 | |
BC138869.1 | - | 10 - 11 | 2233 | 8861891 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
