Assay Details
Gene Symbol: Igf1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010513.2 | NP_034643.2 | 12 - 13 | 3222 | 82 | |
GenBank mRNA | AF056187.1 | - | 12 - 13 | 2620 | 82 | |
AK137220.1 | - | 7 - 8 | 1351 | 82 | ||
AK155884.1 | - | 11 - 12 | 2118 | 82 | ||
BC138868.1 | - | 12 - 13 | 2652 | 8861890 | 82 | |
BC138869.1 | - | 12 - 13 | 2652 | 8861891 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
