Assay Details
Gene Symbol: Ogdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252282.1 | NP_001239211.1 | 11 - 12 | 1500 | 60 | |
NM_001252283.1 | NP_001239212.1 | 11 - 12 | 1488 | 60 | ||
NM_001252287.1 | NP_001239216.1 | 10 - 11 | 1531 | 60 | ||
NM_001252288.1 | NP_001239217.1 | 10 - 11 | 1503 | 60 | ||
NM_010956.4 | NP_035086.2 | 10 - 11 | 1455 | 60 | ||
GenBank mRNA | AK147289.1 | - | 10 - 11 | 1456 | 60 | |
AK150009.1 | - | 10 - 11 | 1421 | 60 | ||
AK169286.1 | - | 10 - 11 | 1447 | 60 | ||
AK220536.1 | - | 11 - 12 | 1462 | 60 | ||
BC013670.1 | - | 10 - 11 | 1389 | 4188988 | 60 | |
BC049104.1 | - | 10 - 11 | 1456 | 6417392 | 60 | |
BC057354.1 | - | 11 - 12 | 1416 | 6827509 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
