Assay Details
Gene Symbol: P4ha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011030.2 | NP_035160.1 | 2 - 3 | 286 | 78 | |
GenBank mRNA | AK030574.1 | - | 2 - 3 | 218 | 78 | |
AK045008.1 | - | 2 - 3 | 215 | 78 | ||
AK149127.1 | - | 2 - 3 | 178 | 78 | ||
AK160798.1 | - | 2 - 3 | 215 | 78 | ||
AK161212.1 | - | 2 - 3 | 219 | 78 | ||
AK169726.1 | - | 2 - 3 | 219 | 78 | ||
BC009654.1 | - | 2 - 3 | 209 | 3484815 | 78 | |
BE985798.1 | - | 2 - 3 | 286 | 78 | ||
BY721363.1 | - | 2 - 3 | 267 | 78 | ||
U16162.1 | - | 1 - 2 | 56 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
