Assay Details
Gene Symbol: P4ha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011030.2 | NP_035160.1 | 5 - 6 | 666 | 81 | |
GenBank mRNA | AK030574.1 | - | 5 - 6 | 598 | 81 | |
AK045008.1 | - | 5 - 6 | 595 | 81 | ||
AK149127.1 | - | 5 - 6 | 558 | 81 | ||
AK160798.1 | - | 5 - 6 | 595 | 81 | ||
AK165468.1 | - | 5 - 6 | 595 | 81 | ||
AK169726.1 | - | 5 - 6 | 599 | 81 | ||
BC009654.1 | - | 5 - 6 | 589 | 3484815 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
