Assay Details
Gene Symbol: Ptprn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011215.2 | NP_035345.2 | 15 - 16 | 2450 | 88 | |
GenBank mRNA | AK138655.1 | - | 15 - 16 | 2442 | 88 | |
AK220255.1 | - | 15 - 16 | 2378 | 88 | ||
BC133678.1 | - | 15 - 16 | 2334 | 40109882 | 88 | |
U57345.1 | - | 15 - 16 | 2337 | 88 | ||
U82439.1 | - | 10 - 11 | 1458 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
