Assay Details
Gene Symbol: Ptprn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011215.2 | NP_035345.2 | 16 - 17 | 2520 | 78 | |
GenBank mRNA | AK138655.1 | - | 16 - 17 | 2512 | 78 | |
BC133678.1 | - | 16 - 17 | 2404 | 40109882 | 78 | |
U57345.1 | - | 16 - 17 | 2407 | 78 | ||
U82439.1 | - | 11 - 12 | 1528 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
