Assay Details
Gene Symbol: Rfx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166414.1 | NP_001159886.1 | 15 - 16 | 1982 | 102 | |
NM_011265.3 | NP_035395.2 | 14 - 15 | 2023 | 102 | ||
GenBank mRNA | AK030008.1 | - | 16 - 17 | 2055 | 102 | |
AK132267.1 | - | 10 - 11 | 1348 | 102 | ||
AK140928.1 | - | 13 - 14 | 1832 | 102 | ||
BC017598.1 | - | 14 - 15 | 1997 | 4483833 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
