Assay Details
Gene Symbol: Ros1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011282.2 | NP_035412.2 | 2 - 3 | 454 | 77 | |
GenBank mRNA | U15443.1 | - | 2 - 3 | 453 | 77 | |
X81650.1 | - | 2 - 3 | 277 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
