Assay Details
Gene Symbol: Ros1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011282.2 | NP_035412.2 | 31 - 32 | 5350 | 74 | |
GenBank mRNA | U15443.1 | - | 31 - 32 | 5349 | 74 | |
X81650.1 | - | 31 - 32 | 5173 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
