Assay Details
Gene Symbol: Ros1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011282.2 | NP_035412.2 | 37 - 38 | 6202 | 65 | |
GenBank mRNA | U15443.1 | - | 37 - 38 | 6201 | 65 | |
X81650.1 | - | 37 - 38 | 6025 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
