Assay Details
Gene Symbol: Ros1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011282.2 | NP_035412.2 | 9 - 10 | 1167 | 134 | |
GenBank mRNA | U15443.1 | - | 9 - 10 | 1166 | 134 | |
X81650.1 | - | 9 - 10 | 990 | 134 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
