Assay Details
Gene Symbol: Map3k1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011945.2 | NP_036075.2 | 11 - 12 | 2095 | 75 | |
GenBank mRNA | AF117340.1 | - | 11 - 12 | 2095 | 75 | |
L13103.1 | - | 1 - 2 | 104 | 75 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the L13103.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
