Assay Details
Gene Symbol: Sema4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013659.4 | NP_038687.2 | 7 - 8 | 952 | 63 | |
GenBank mRNA | AK028896.1 | - | 7 - 8 | 915 | 63 | |
AK030331.1 | - | 6 - 7 | 952 | 63 | ||
AK134973.1 | - | 6 - 7 | 883 | 63 | ||
AK145570.1 | - | 7 - 8 | 953 | 63 | ||
AK154969.1 | - | 6 - 7 | 733 | 63 | ||
AK173249.1 | - | 7 - 8 | 900 | 63 | ||
DQ073331.1 | - | 6 - 7 | 708 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
