Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500015210.mm.1 |
TC1500001934.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500015210.mm.1 |
NM_001282040.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015210.mm.1 |
NM_001282041.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015210.mm.1 |
NM_013872.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015202.mm.1 |
TC1500001934.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500015202.mm.1 |
NM_001282040.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015202.mm.1 |
NM_001282041.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015202.mm.1 |
NM_013872.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015211.mm.1 |
TC1500001934.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500015211.mm.1 |
NM_001282040.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015211.mm.1 |
NM_001282041.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015211.mm.1 |
NM_013872.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015213.mm.1 |
TC1500001934.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500015213.mm.1 |
NM_001282040.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015213.mm.1 |
NM_001282041.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015213.mm.1 |
NM_013872.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015208.mm.1 |
TC1500001934.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500015208.mm.1 |
NM_001282041.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015208.mm.1 |
NM_013872.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015207.mm.1 |
TC1500001934.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500015207.mm.1 |
NM_013872.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015215.mm.1 |
TC1500001934.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500015215.mm.1 |
NM_001282040.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015215.mm.1 |
NM_001282041.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015215.mm.1 |
NM_013872.4 |
Refseq |