Assay Details
Gene Symbol: Slc12a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020333.2 | NP_065066.2 | 3 - 4 | 695 | 96 | |
GenBank mRNA | AF332063.1 | - | 3 - 4 | 365 | 96 | |
AF332064.1 | - | 3 - 4 | 365 | 96 | ||
AK122460.1 | - | 3 - 4 | 429 | 96 | ||
AK147262.1 | - | 3 - 4 | 697 | 96 | ||
AK165769.1 | - | 3 - 4 | 443 | 96 | ||
BC054808.1 | - | 3 - 4 | 361 | 6838880 | 96 | |
KJ535320.1 | - | 3 - 4 | 386 | 96 | ||
KJ535321.1 | - | 3 - 4 | 386 | 96 | ||
KJ535322.1 | - | 3 - 4 | 542 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
