Assay Details
Gene Symbol: Slc12a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020333.2 | NP_065066.2 | 5 - 6 | 895 | 76 | |
GenBank mRNA | AF332063.1 | - | 5 - 6 | 565 | 76 | |
AF332064.1 | - | 5 - 6 | 565 | 76 | ||
AK122460.1 | - | 5 - 6 | 629 | 76 | ||
AK147262.1 | - | 5 - 6 | 897 | 76 | ||
AK165769.1 | - | 5 - 6 | 643 | 76 | ||
BC054808.1 | - | 5 - 6 | 561 | 6838880 | 76 | |
KJ535320.1 | - | 5 - 6 | 586 | 76 | ||
KJ535321.1 | - | 5 - 6 | 586 | 76 | ||
KJ535322.1 | - | 5 - 6 | 742 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
