Assay Details
Gene Symbol: Hadha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178878.2 | NP_849209.1 | 4 - 5 | 347 | 106 | |
GenBank mRNA | AK029017.1 | - | 4 - 5 | 348 | 106 | |
AK035399.1 | - | 4 - 5 | 330 | 106 | ||
AK150188.1 | - | 4 - 5 | 345 | 106 | ||
AK170478.1 | - | 4 - 5 | 348 | 106 | ||
AK170683.1 | - | 4 - 5 | 348 | 106 | ||
AK171469.1 | - | 4 - 5 | 348 | 106 | ||
AK183745.1 | - | 3 - 4 | 225 | 106 | ||
AK213006.1 | - | 3 - 4 | 225 | 106 | ||
AK218789.1 | - | 3 - 4 | 224 | 106 | ||
BC037009.1 | - | 4 - 5 | 335 | 5134605 | 106 | |
BC046978.1 | - | 4 - 5 | 321 | 6315204 | 106 | |
BC058569.1 | - | 4 - 5 | 343 | 6468514 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
