Assay Details
Gene Symbol: Zfp598 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183149.1 | NP_898972.1 | 11 - 12 | 2377 | 74 | |
GenBank mRNA | AK131127.1 | - | 12 - 13 | 2312 | 74 | |
AK138860.1 | - | 8 - 9 | 2490 | 74 | ||
BC023040.1 | - | 4 - 5 | 279 | 74 | ||
BC024690.1 | - | 11 - 12 | 2162 | 74 | ||
BC038138.1 | - | 1 - 2 | 139 | 74 | ||
BC050859.1 | - | 11 - 12 | 2377 | 6328878 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31098877 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31098878 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31098880 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31098881 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31098882 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2051894 | BC050859.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2051894 | BC038138.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2051894 | AK136995.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2051894 | AK213857.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2051894 | BC023040.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 511879 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 465707 | AK031035.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 822601 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 231048 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 837785 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 371685 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 117489 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 538238 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 110830 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P111057 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P143740 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2051894 | BC050859.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2051894 | BC038138.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2051894 | AK136995.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2051894 | AK213857.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2051894 | BC023040.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P329855 | BC050859.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P329855 | BC024690.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P329855 | BC038138.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P329855 | AK136995.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P329855 | AK138860.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31098896 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31098861 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31098872 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31098880 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31098875 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31098879 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31098862 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31098888 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31098878 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31098892 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31098890 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31098885 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31098891 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31098852 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31098856 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31098858 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31098876 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31098863 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31098884 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31098854 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31098882 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31098893 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31098867 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31098869 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31098865 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P143740 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P111057 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08740245 | - | - |
Whole Mouse Genome G4122A | A_51_P329855 | BC050859.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P329855 | BC024690.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P329855 | BC038138.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P329855 | AK136995.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P329855 | AK138860.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122832 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P143740 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P111057 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13344598 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13270003 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13497044 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13149887 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13270000 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31098880 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31098878 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31098882 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31098881 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31098877 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098854 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098880 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098836 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098995 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098956 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098917 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098872 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099006 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098934 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098921 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098897 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098861 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099021 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098907 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098981 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098833 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098968 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098957 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099023 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099019 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098856 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098911 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098914 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098960 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098888 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098989 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098944 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098899 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098882 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098947 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098945 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098884 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098840 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098875 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098958 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098919 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098928 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098867 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098902 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098987 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098923 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098860 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099024 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098961 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099000 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098885 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098959 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098969 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098992 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098982 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098977 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098843 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098954 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099009 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099007 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099017 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098996 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099011 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098926 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098890 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098946 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098893 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098865 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098975 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098891 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098913 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098835 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098984 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098896 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098936 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098877 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099029 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099015 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098918 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098950 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098905 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098949 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099008 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098986 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098967 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098894 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099002 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098939 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098971 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098842 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098924 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098966 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098838 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098964 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098831 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098941 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098910 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098879 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099012 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098962 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098857 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098869 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098930 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099013 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098932 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098980 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098938 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098827 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098983 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098922 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098832 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098875 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098869 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098923 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13568703 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098926 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098921 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098914 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13118162 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13149886 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13319342 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098885 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098916 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13282997 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098910 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098912 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098899 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098901 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13497049 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13388298 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098861 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098856 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098932 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13497044 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098882 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098907 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13427696 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13104913 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13251198 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098854 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098867 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098913 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098918 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13270004 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13427681 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13388300 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098876 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098897 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098872 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098905 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098924 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098900 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098863 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13118159 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13427711 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098888 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098919 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098865 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098975 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098870 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098860 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098890 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10442553 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10442554 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10442555 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10442561 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10442552 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10442558 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10442557 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10442551 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10442562 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10442563 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10442560 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10442559 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10442556 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10442550 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10442564 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1460691_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10442549 | - | - |
Mouse Genome 430 2.0 Array | 1460691_at | - | - |
Mouse Genome 430 2.0 Array | 1460691_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 213753_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 213753_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1700002656.mm.1 | TC1700000347.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700002656.mm.1 | NM_183149.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700002655.mm.1 | TC1700000347.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700002655.mm.1 | NM_183149.2 | Refseq |
Mouse Expression Set 430 | 1460691_at | - | - |
Mouse Expression Set 430 | 1460691_PM_at | - | - |
Murine Genome U74v2 Set | 100482_at | - | - |
Murine Genome U74v2 Set | 100482_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1460691_at | - | - |
Mouse Genome 430A 2.0 Array | 1460691_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1460691_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1460691_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2900156 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2900156 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002900156 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1257200 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1257200 | - | - |
mouse-6 v1.1 (Illumina) | scl51009.12.1_25-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2900156 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl51009.12.1_25-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2900156 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1257200 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1257200 | - | - |
