Assay Details
Gene Symbol: Zfp598 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183149.1 | NP_898972.1 | 4 - 5 | 681 | 94 | |
GenBank mRNA | AK131127.1 | - | 4 - 5 | 700 | 94 | |
AK138860.1 | - | 2 - 3 | 260 | 94 | ||
BC024690.1 | - | 4 - 5 | 475 | 94 | ||
BC050859.1 | - | 4 - 5 | 681 | 6328878 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
