Assay Details
Gene Symbol: Zfp598 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183149.1 | NP_898972.1 | 7 - 8 | 1199 | 128 | |
GenBank mRNA | AK131127.1 | - | 7 - 8 | 1218 | 128 | |
AK138860.1 | - | 4 - 5 | 1312 | 128 | ||
BC024690.1 | - | 7 - 8 | 984 | 128 | ||
BC050859.1 | - | 7 - 8 | 1199 | 6328878 | 128 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
