Assay Details
Gene Symbol: Akap6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198111.2 | NP_932779.2 | 3 - 4 | 677 | 75 | |
GenBank mRNA | AK147248.1 | - | 3 - 4 | 678 | 75 | |
AK147371.1 | - | 3 - 4 | 678 | 75 | ||
BC057114.1 | - | 2 - 3 | 627 | 6405025 | 75 | |
DQ233645.1 | - | 3 - 4 | 730 | 75 | ||
DQ233646.1 | - | 2 - 3 | 524 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the DQ233646.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
