Assay Details
Gene Symbol: Akap6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198111.2 | NP_932779.2 | 5 - 6 | 2550 | 100 | |
GenBank mRNA | AK147248.1 | - | 5 - 6 | 2551 | 100 | |
AK147371.1 | - | 5 - 6 | 2551 | 100 | ||
BC057114.1 | - | 4 - 5 | 2500 | 6405025 | 100 | |
DQ233645.1 | - | 5 - 6 | 2606 | 100 | ||
DQ233646.1 | - | 4 - 5 | 2400 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
