Assay Details
Gene Symbol: Akap6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198111.2 | NP_932779.2 | 7 - 8 | 2821 | 105 | |
GenBank mRNA | AK147248.1 | - | 7 - 8 | 2822 | 105 | |
AK147371.1 | - | 7 - 8 | 2822 | 105 | ||
BC057114.1 | - | 6 - 7 | 2771 | 6405025 | 105 | |
DQ233645.1 | - | 7 - 8 | 2877 | 105 | ||
DQ233646.1 | - | 6 - 7 | 2671 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
