Assay Details
Gene Symbol: Akap6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198111.2 | NP_932779.2 | 9 - 10 | 3087 | 85 | |
GenBank mRNA | AK147248.1 | - | 9 - 10 | 3088 | 85 | |
AK147371.1 | - | 9 - 10 | 3088 | 85 | ||
BC057114.1 | - | 8 - 9 | 3037 | 6405025 | 85 | |
DQ233645.1 | - | 9 - 10 | 3143 | 85 | ||
DQ233646.1 | - | 8 - 9 | 2937 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
