Assay Details
Gene Symbol: Pfn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011072.4 | NP_035202.1 | 1 - 2 | 306 | 58 | |
GenBank mRNA | AK011066.1 | - | 1 - 2 | 291 | 58 | |
AK028117.1 | - | 1 - 2 | 257 | 58 | ||
AK150780.1 | - | 1 - 2 | 263 | 58 | ||
AK152491.1 | - | 1 - 2 | 292 | 58 | ||
AK160191.1 | - | 1 - 2 | 292 | 58 | ||
AV448567.1 | - | 1 - 2 | 307 | 58 | ||
BC002080.1 | - | 1 - 2 | 279 | 3490976 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
