Assay Details
Gene Symbol: Rad23a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297606.1 | NP_001284535.1 | 1 - 2 | 170 | 158 | |
NM_001297607.1 | NP_001284536.1 | 1 - 2 | 170 | 158 | ||
NM_009010.5 | NP_033036.2 | 1 - 2 | 170 | 158 | ||
GenBank mRNA | AK038300.1 | - | 1 - 2 | 147 | 158 | |
AK045224.1 | - | 1 - 2 | 160 | 158 | ||
AK161875.1 | - | 1 - 2 | 147 | 158 | ||
AK165467.1 | - | 1 - 2 | 171 | 158 | ||
BC003846.1 | - | 1 - 2 | 158 | 158 | ||
BC138684.1 | - | 1 - 2 | 130 | 8861705 | 158 | |
BC145372.1 | - | 1 - 2 | 130 | 9053930 | 158 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
