Assay Details
Gene Symbol: Tuba3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009446.2 | NP_033472.1 | 4 - 5 | 1124 | 142 | |
GenBank mRNA | AK132775.1 | - | 4 - 5 | 1124 | 142 | |
BC050770.1 | - | 4 - 5 | 1110 | 6774775 | 142 | |
BC089547.1 | - | 4 - 5 | 1110 | 6771690 | 142 | |
M13442.1 | - | 4 - 5 | 1112 | 142 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24819776 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24819777 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24819778 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1979126 | NM_009446.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1979126 | BC089547.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2015375 | M13442.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2015375 | NM_009446.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2015375 | BC089547.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 228726 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 39001 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 249277 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 380715 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 515811 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 12832 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P119432 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2015375 | M13442.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2015375 | NM_009446.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2015375 | BC089547.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1979126 | NM_009446.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1979126 | BC089547.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P145402 | M13442.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P145402 | NM_009446.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P145402 | AK132775.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P145402 | BC050770.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P145402 | BC089547.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P145404 | M13442.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P145404 | NM_009446.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P145404 | AK132775.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P145404 | BC050770.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P145404 | BC089547.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24819769 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24819771 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24819792 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24819780 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24819791 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24819793 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24819776 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24819795 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24819764 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24819782 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24819773 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24819775 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24819774 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24819794 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24819783 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24819785 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24819778 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24819765 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24819768 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24819787 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24819789 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24819781 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06057799 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P119432 | - | - |
Whole Mouse Genome G4122A | A_51_P145404 | M13442.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P145404 | NM_009446.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P145404 | AK132775.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P145404 | BC050770.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P145404 | BC089547.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P145402 | M13442.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P145402 | NM_009446.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P145402 | AK132775.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P145402 | BC050770.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P145402 | BC089547.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117232 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P119432 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05162924 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05162923 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05162922 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05812613 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05162917 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24819776 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24819778 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24819777 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819722 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819692 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819680 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819706 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819690 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819715 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819774 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819780 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819760 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819755 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819708 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819792 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819773 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819795 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819678 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819686 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819778 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819699 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819720 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819771 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819679 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819724 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819681 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819677 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819802 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819753 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819775 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819709 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819785 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819693 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819705 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819717 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819683 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819700 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819732 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819764 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819702 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819789 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819735 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819739 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819703 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819684 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819737 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819800 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819689 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819787 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819782 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819799 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819769 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819762 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819768 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819687 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819755 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819695 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819771 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819759 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819792 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819689 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819770 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05507478 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05105316 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819692 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819798 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819784 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819682 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819789 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819677 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819775 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819739 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819731 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819743 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05162930 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819730 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819796 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819785 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819778 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819793 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819774 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819782 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819686 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819794 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05340494 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819741 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819680 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819742 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819776 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819795 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05808794 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819773 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819687 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819762 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05088996 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819754 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05812613 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819797 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05340495 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819737 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819751 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819696 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819735 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05268831 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819681 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819684 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819690 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819678 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819791 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819787 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819679 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819760 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24819764 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10547998 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10547997 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10547999 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10547996 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1448296_PM_x_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10547995 | - | - |
Mouse Genome 430 2.0 Array | 1448296_PM_x_at | - | - |
Mouse Genome 430 2.0 Array | 1416311_PM_s_at | - | - |
Mouse Genome 430 2.0 Array | 1416311_s_at | - | - |
Mouse Genome 430 2.0 Array | 1448296_x_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22144_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22144_at | - | - |
Affymetrix Murine Genome 6500A Array | Msa12580_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0600024564.mm.1 | TC0600003144.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600024564.mm.1 | NM_009446.3 | Refseq |
Mouse Expression Set 430 | 1448296_PM_x_at | - | - |
Mouse Expression Set 430 | 1416311_s_at | - | - |
Mouse Expression Set 430 | 1416311_PM_s_at | - | - |
Mouse Expression Set 430 | 1448296_x_at | - | - |
Murine Genome U74v2 Set | 160420_PM_r_at | - | - |
Murine Genome U74v2 Set | 160420_r_at | - | - |
Murine Genome U74v2 Set | 93924_f_at | - | - |
Murine Genome U74v2 Set | 93924_PM_f_at | - | - |
Murine Genome U74v2 Set | 167535_PM_at | - | - |
Murine Genome U74v2 Set | 167535_at | - | - |
Mouse Genome 430A 2.0 Array | 1448296_PM_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1448296_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1416311_PM_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1416311_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 4540347 | - | - |
MouseWG-6 v2 Expression BeadChip | 610128 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6980181 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 4540347 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 610128 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6420070 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000610128 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004540347 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006980181 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006420070 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2491565 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2505467 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2467287 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1220219 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1220219 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2505467 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2491565 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2467287 | - | - |
mouse-6 v1.1 (Illumina) | scl0001181.1_4-S | - | - |
mouse-6 v1.1 (Illumina) | scl0022144.2_97-S | - | - |
mouse-6 v1.1 (Illumina) | scl022144.4_75-S | - | - |
mouse-6 v1.1 (Illumina) | scl0022144.2_2-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4540347 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 610128 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6420070 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6980181 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0022144.2_97-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0022144.2_2-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl022144.4_75-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0001181.1_4-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4540347 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6980181 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 610128 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6420070 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1220219 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2491565 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2505467 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2467287 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2491565 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2467287 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1220219 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2505467 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0022144.2_97-S | - | - |
