Assay Details
Gene Symbol: Cox7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025379.2 | NP_079655.1 | 1 - 2 | 175 | 112 | |
GenBank mRNA | AK002593.1 | - | 1 - 2 | 122 | 112 | |
AK003414.1 | - | 1 - 2 | 126 | 112 | ||
AK132086.1 | - | 1 - 2 | 171 | 112 | ||
BC024350.1 | - | 1 - 2 | 116 | 4987472 | 112 | |
BC089538.1 | - | 1 - 2 | 175 | 6533206 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P160664 | NM_025379.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P160664 | AK002593.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P160664 | AK003414.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P160664 | AK132086.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P160664 | BC024350.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P160664 | NM_025379.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P160664 | AK002593.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P160664 | AK003414.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P160664 | AK132086.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P160664 | BC024350.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 133229 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924443 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 971392 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 319455 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 131618 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 125453 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P129341 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P160664 | NM_025379.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P160664 | AK002593.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P160664 | AK003414.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P160664 | AK132086.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P160664 | BC024350.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P160664 | NM_025379.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P160664 | AK002593.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P160664 | AK003414.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P160664 | AK132086.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P160664 | BC024350.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P160664 | NM_025379.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P160664 | AK002593.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P160664 | AK003414.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P160664 | AK132086.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P160664 | BC024350.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P160664 | NM_025379.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P160664 | AK002593.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P160664 | AK003414.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P160664 | AK132086.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P160664 | BC024350.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32582429 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32582463 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32582435 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32582434 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32582464 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32582426 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32582437 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32582428 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32582425 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32582430 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32582461 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32582424 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32582433 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32582432 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P129341 | - | - |
Whole Mouse Genome G4122A | A_51_P160664 | NM_025379.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P160664 | AK002593.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P160664 | AK003414.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P160664 | AK132086.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P160664 | BC024350.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P160664 | NM_025379.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P160664 | AK002593.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P160664 | AK003414.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P160664 | AK132086.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P160664 | BC024350.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P129341 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14982418 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582393 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582457 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582507 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582495 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582407 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582473 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582513 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582480 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582497 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582509 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582458 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582488 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582481 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582511 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582498 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582452 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582486 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582491 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582412 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582437 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582432 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582417 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582504 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582404 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582399 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582379 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582381 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582479 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582420 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582405 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582418 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582463 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582476 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582485 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582499 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582483 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582490 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582496 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582425 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582401 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582508 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582443 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582397 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582395 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582501 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582409 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582475 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582430 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582389 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582439 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582454 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582434 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582516 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582449 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582482 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582426 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582489 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582459 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582441 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582414 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582390 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582493 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582392 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582461 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582447 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582387 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582411 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582445 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582451 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582440 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582470 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582383 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582429 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582424 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582438 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582464 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32582398 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32582418 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32582438 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32582445 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32582461 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32582421 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32582449 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32582432 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32582464 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32582439 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32582440 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32582422 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14700132 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32582443 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32582454 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32582457 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32582428 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32582430 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32582420 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32582424 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32582458 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32582437 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32582426 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32582434 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32582429 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32582433 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14787427 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32582451 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32582459 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32582447 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32582441 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32582452 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10601358 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10358710 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10601359 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10601357 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10601356 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10358709 | - | - |
Mouse Genome 430 2.0 Array | 1443944_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1443944_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0X00007017.mm.1 | TC0X00001065.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0X00007017.mm.1 | NM_025379.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00007017.mm.1 | NM_025379.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00007011.mm.1 | TC0X00001065.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0X00007011.mm.1 | NM_025379.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00007011.mm.1 | NM_025379.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00007020.mm.1 | TC0X00001065.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0X00007020.mm.1 | NM_025379.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00007020.mm.1 | NM_025379.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00007018.mm.1 | TC0X00001065.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0X00007018.mm.1 | NM_025379.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00007018.mm.1 | NM_025379.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00007012.mm.1 | TC0X00001065.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0X00007012.mm.1 | NM_025379.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00007012.mm.1 | NM_025379.2 | Refseq |
Murine Genome U74v2 Set | 101580_PM_at | - | - |
Murine Genome U74v2 Set | 101580_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 894223 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 102570041 | - | - |
MouseWG-6 v2 Expression BeadChip | 2340504 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 102570041 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2340504 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002340504 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2671689 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2671689 | - | - |
mouse-6 v1.1 (Illumina) | scl0066142.1_255-S | - | - |
mouse-6 v1.1 (Illumina) | scl25070.6.1_66-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 102570041 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2340504 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0066142.1_255-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102570041 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2340504 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2671689 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2671689 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0066142.1_255-S | - | - |
