Assay Details
Gene Symbol: Uhrf1bp1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029166.2 | NP_083442.2 | 13 - 14 | 2004 | 80 | |
GenBank mRNA | AB093261.1 | - | 13 - 14 | 1969 | 80 | |
AK087280.1 | - | 11 - 12 | 1556 | 80 | ||
AK141050.1 | - | 13 - 14 | 2013 | 80 | ||
BC079645.1 | - | 5 - 6 | 673 | 80 | ||
BC132134.1 | - | 13 - 14 | 1797 | 40130411 | 80 | |
BC138092.1 | - | 13 - 14 | 1797 | 8861111 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
