Assay Details
Gene Symbol: Cdc42ep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027219.3 | NP_081495.1 | 2 - 3 | 804 | 66 | |
GenBank mRNA | AK007896.1 | - | 2 - 3 | 805 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30362681 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30362677 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30362719 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30362679 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30362718 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30362680 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30362678 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30362720 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30362721 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30362683 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P573255 | NM_027219.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P573255 | AK007896.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P573255 | NM_027219.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P573255 | AK007896.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 179693 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 212025 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 356454 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 834781 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 123369 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P123542 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P573255 | NM_027219.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P573255 | AK007896.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P573255 | NM_027219.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P573255 | AK007896.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P180629 | BC083130.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P180629 | BC016250.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P180629 | NM_027219.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P180629 | AK007896.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362700 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362660 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362697 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362682 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362681 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362651 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362687 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362673 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362695 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362662 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362684 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362665 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362696 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362676 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362667 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362691 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362677 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362671 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362689 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362702 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362669 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362657 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362650 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362688 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362654 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362672 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362679 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362674 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362683 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362699 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362694 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03077810 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P123542 | - | - |
Whole Mouse Genome G4122A | A_51_P180629 | BC083130.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P180629 | BC016250.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P180629 | NM_027219.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P180629 | AK007896.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P573255 | NM_027219.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P573255 | AK007896.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P573255 | NM_027219.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P573255 | AK007896.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115519 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P123542 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12175355 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12250970 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11836981 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12129893 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11836982 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30362719 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30362721 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30362677 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30362720 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30362683 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30362680 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30362718 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30362678 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30362679 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30362681 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362695 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362791 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362794 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362754 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362765 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362688 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362785 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362786 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362706 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362676 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362684 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362764 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362721 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362710 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362780 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362634 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362660 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362642 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362742 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362696 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362646 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362669 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362768 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362747 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362744 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362790 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362628 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362749 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362756 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362643 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362630 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362658 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362638 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362641 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362739 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362737 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362777 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362672 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362687 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362763 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362732 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362797 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362691 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362665 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362631 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362751 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362616 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362723 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362699 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362703 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362618 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362760 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362717 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362700 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362655 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362740 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362626 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362724 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362662 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362639 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362715 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362741 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362681 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362766 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362614 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362623 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362689 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362735 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362752 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362636 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362767 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362726 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362773 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362632 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362776 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362729 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362799 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362673 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362759 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362792 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362800 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362708 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362758 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362644 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362679 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362677 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362775 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362632 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362658 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362646 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362638 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362643 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362667 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362706 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12331502 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362628 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362662 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362721 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11855459 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362732 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362696 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12129892 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362631 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362660 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362677 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362742 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362681 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362648 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362620 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11975332 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362688 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362669 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362726 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362655 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12130769 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362639 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362739 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11855470 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362699 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362630 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362674 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362729 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12174658 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362704 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362735 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362644 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362703 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362671 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362618 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362626 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362616 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362665 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362672 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362617 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362666 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362715 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362740 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362714 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362642 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362651 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362737 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362695 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362725 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11975341 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12084060 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362745 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12130764 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362621 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362650 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12331498 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11975338 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362691 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362623 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362675 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362741 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362676 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362710 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362652 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12130768 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362744 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30362687 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10425117 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10425119 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10425118 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1424376_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10425116 | - | - |
Mouse Genome 430 2.0 Array | 1424376_at | - | - |
Mouse Genome 430 2.0 Array | 1442512_at | - | - |
Mouse Genome 430 2.0 Array | 1442512_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1424376_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 104445_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 104445_PM_at | - | - |
Mouse Expression Set 430 | 1424376_at | - | - |
Mouse Expression Set 430 | 1424376_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1424376_at | - | - |
Mouse Genome 430A 2.0 Array | 1424376_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1424376_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1424376_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 680041 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 6550279 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6550279 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006550279 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1250030 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2523947 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1250030 | - | - |
mouse-6 v1.1 (Illumina) | scl47764.3_554-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6550279 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl47764.3_554-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6550279 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1250030 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1250030 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl47764.3_554-S | - | - |
