Assay Details
Gene Symbol: Grin3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033351.2 | NP_001028523.1 | 1 - 2 | 1386 | 92 | |
NM_001276355.1 | NP_001263284.1 | 1 - 2 | 1386 | 92 | ||
GenBank mRNA | AK044171.1 | - | 1 - 2 | 1363 | 92 | |
AK138366.1 | - | 1 - 2 | 1387 | 92 | ||
AK173314.1 | - | 1 - 2 | 387 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
