Assay Details
Gene Symbol: Arhgef17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081116.1 | NP_001074585.1 | 13 - 14 | 5096 | 106 | |
GenBank mRNA | AK122250.1 | - | 13 - 14 | 1751 | 106 | |
BC035332.1 | - | 4 - 5 | 571 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
