Assay Details
Gene Symbol: Arhgef17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081116.1 | NP_001074585.1 | 16 - 17 | 5732 | 91 | |
GenBank mRNA | AK122250.1 | - | 16 - 17 | 2387 | 91 | |
BC028887.1 | - | 3 - 4 | 351 | 91 | ||
BC035332.1 | - | 7 - 8 | 1207 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
