Assay Details
Gene Symbol: Cdc25c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK141737.1 | - | 9 - 10 | 1033 | 80 | |
AK168287.1 | - | 9 - 10 | 1028 | 80 | ||
BC004669.1 | - | 8 - 9 | 737 | 3498846 | 80 | |
L16926.1 | - | 8 - 9 | 1215 | 80 | ||
U15562.1 | - | 9 - 10 | 999 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
