Assay Details
Gene Symbol: Rsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081267.2 | NP_001074736.1 | 11 - 12 | 3079 | 123 | |
GenBank mRNA | BC054454.1 | - | 6 - 7 | 569 | 123 | |
BC085127.1 | - | 10 - 11 | 4008 | 123 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
