Assay Details
Gene Symbol: Tbce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK082940.1 | - | 8 - 9 | 872 | 82 | |
AK167383.1 | - | 8 - 9 | 860 | 82 | ||
AK167848.1 | - | 8 - 9 | 856 | 82 | ||
AY082332.1 | - | 8 - 9 | 826 | 82 | ||
BC050206.1 | - | 8 - 9 | 834 | 30009463 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
