Assay Details
Gene Symbol: Ptpn12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011203.2 | NP_035333.2 | 9 - 10 | 813 | 96 | |
GenBank mRNA | AK154107.1 | - | 9 - 10 | 905 | 96 | |
BC051980.1 | - | 9 - 10 | 813 | 6402545 | 96 | |
X63440.1 | - | 9 - 10 | 814 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
