Assay Details
Gene Symbol: Hgf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289458.1 | NP_001276387.1 | 5 - 6 | 684 | 68 | |
NM_001289459.1 | NP_001276388.1 | 5 - 6 | 673 | 68 | ||
NM_001289460.1 | NP_001276389.1 | 5 - 6 | 684 | 68 | ||
NM_010427.5 | NP_034557.3 | 5 - 6 | 656 | 68 | ||
GenBank mRNA | AF042856.1 | - | 4 - 5 | 485 | 68 | |
AK042121.1 | - | 5 - 6 | 602 | 68 | ||
AK052289.1 | - | 5 - 6 | 685 | 68 | ||
BC119228.1 | - | 5 - 6 | 543 | 8733977 | 68 | |
BC144764.1 | - | 5 - 6 | 555 | 68 | ||
D10212.1 | - | 4 - 5 | 479 | 68 | ||
S71816.1 | - | 4 - 5 | 479 | 68 | ||
X72307.1 | - | 4 - 5 | 540 | 68 | ||
X84046.1 | - | 4 - 5 | 519 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
