Assay Details
Gene Symbol: Sema3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013657.5 | NP_038685.3 | 13 - 14 | 1628 | 140 | |
GenBank mRNA | AK034239.1 | - | 13 - 14 | 1687 | 140 | |
AK077231.1 | - | 2 - 3 | 751 | 140 | ||
AK140348.1 | - | 2164 | 140 | |||
BC066852.1 | - | 13 - 14 | 1628 | 30093230 | 140 | |
X85994.1 | - | 13 - 14 | 1580 | 140 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
