Assay Details
Gene Symbol: Gpld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008156.2 | NP_032182.2 | 12 - 13 | 1203 | 138 | |
GenBank mRNA | AF050666.1 | - | 12 - 13 | 1145 | 138 | |
AK004957.2 | - | 11 - 12 | 907 | 138 | ||
AY081194.1 | - | 11 - 12 | 930 | 138 | ||
AY333960.1 | - | 11 - 12 | 887 | 138 | ||
BC019146.1 | - | 11 - 12 | 916 | 5052822 | 138 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
