Assay Details
Gene Symbol: Slc37a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK210256.1 | - | 1 - 2 | 89 | 70 | |
AK210278.1 | - | 1 - 2 | 89 | 70 | ||
BC100391.1 | - | 1 - 2 | 84 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
