Assay Details
Gene Symbol: Ppfibp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163557.1 | NP_001157029.1 | 16 - 17 | 1623 | 77 | |
NM_008905.2 | NP_032931.2 | 17 - 18 | 1892 | 77 | ||
GenBank mRNA | AK012152.1 | - | 13 - 14 | 1282 | 77 | |
AK166921.1 | - | 7 - 8 | 760 | 77 | ||
BC003966.1 | - | 2 - 3 | 192 | 77 | ||
BC055935.1 | - | 6 - 7 | 493 | 77 | ||
BC100346.1 | - | 16 - 17 | 1623 | 30936010 | 77 | |
U79024.1 | - | 17 - 18 | 1677 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
