Assay Details
Gene Symbol: Kat8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026370.1 | NP_080646.1 | 4 - 5 | 534 | 74 | |
GenBank mRNA | AK003264.1 | - | 4 - 5 | 534 | 74 | |
AK018002.1 | - | 4 - 5 | 531 | 74 | ||
AK132000.1 | - | 4 - 5 | 535 | 74 | ||
AK146710.1 | - | 4 - 5 | 536 | 74 | ||
BC036284.1 | - | 4 - 5 | 542 | 5136304 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
