Assay Details
Gene Symbol: Inpp5f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178641.5 | NP_848756.2 | 6 - 7 | 901 | 93 | |
GenBank mRNA | AK030870.1 | - | 6 - 7 | 903 | 93 | |
AK039468.1 | - | 5 - 6 | 899 | 93 | ||
AK129249.1 | - | 6 - 7 | 858 | 93 | ||
AK150418.1 | - | 6 - 7 | 941 | 93 | ||
AK154996.1 | - | 6 - 7 | 906 | 93 | ||
BC125437.1 | - | 6 - 7 | 789 | 40129952 | 93 | |
BC137700.1 | - | 6 - 7 | 789 | 8860719 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
