Assay Details
Gene Symbol: E2f3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010093.3 | NP_034223.1 | 2 - 3 | 874 | 84 | |
GenBank mRNA | AF015948.1 | - | 2 - 3 | 176 | 84 | |
AK133828.1 | - | 2 - 3 | 876 | 84 | ||
AK154168.1 | - | 2 - 3 | 316 | 84 | ||
BC059262.1 | - | 2 - 3 | 273 | 84 | ||
BU705601.1 | - | 2 - 3 | 274 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
