Assay Details
Gene Symbol: Sidt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289668.1 | NP_001276597.1 | 11 - 12 | 1424 | 73 | |
NM_172257.4 | NP_758461.1 | 11 - 12 | 1424 | 73 | ||
GenBank mRNA | AK155875.1 | - | 11 - 12 | 1328 | 73 | |
BC023957.1 | - | 11 - 12 | 1428 | 5321190 | 73 | |
BC051101.1 | - | 11 - 12 | 1152 | 5052849 | 73 | |
BQ179223.1 | - | 4 - 5 | 263 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
